Cadmium(II) telluride ≥99.999% (Cd basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Cadmium telluride

48-2000-5G 48-2000-25G
100211-130EA 94.83 USD
100211-130 100201-002
Cadmium(II) telluride ≥99.999% (Cd basis), PURATREM
Cadmium (II) telluride
Formula: CdTe
Melting Pt: 1041 °C
Density: 6.20
Storage Temperature: Ambient
MDL Number: MFCD00015998
CAS Number: 1306-25-8
EINECS: 215-149-9
UN: 2570
ADR: 6.1,III
Merck Index: 12,01667

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR