Tetraamminepalladium(II) tetrachloropalladate(II) ≥99%

Supplier: STREM CHEMICALS INC MS

46-2143-5G 46-2143-1G
100197-158EA 527.94 USD
100197-158 100200-998
Tetraamminepalladium(II) tetrachloropalladate(II) ≥99%
Tetraamminepalladium(II) tetrachloropalladate(II)
Formula: [Pd(NH₃)₄][PdCl₄]
MDL Number: MFCD00049783
CAS Number: 13820-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR