Zirconium ≥99.8%, foil

Supplier: STREM CHEMICALS INC MS

40-0070-50X50MM 40-0070-100X425MM 40-0070-100X100MM
100204-046EA 106.15 USD
100204-046 100196-944 100200-936
Zirconium ≥99.8%, foil
Zirconium
Formula: Zr
Boiling Pt: 3577 °C
Melting Pt: 1852 °C
Density: 6,506
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR