Praseodymium ≥99.9%, ingot

Supplier: STREM CHEMICALS INC MS
Warning

93-5942-50G 93-5942-10G
100200-778EA 961.05 USD
100200-778 100200-776
Praseodymium ≥99.9%, ingot
Praseodymium
Formula: Pr
MW: 140.91
Boiling Pt: 3127 °C
Melting Pt: 935 °C
Density: 6.782
MDL Number: MFCD00011174
CAS Number: 7440-10-0
EINECS: 231-120-3
UN: 2813
ADR: 4.3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR