Yttrium(III) carbonate trihydrate ≥99.9% (Y basis)

Supplier: STREM CHEMICALS INC MS

93-3934-25G 93-3934-100G
100209-112EA 92 USD
100209-112 100200-758
Yttrium(III) carbonate trihydrate ≥99.9% (Y basis)
Yttrium(III) carbonate trihydrate
MDL Number: MFCD00149944
CAS Number: 5970-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR