Strontium oxide ≥99.9%

Supplier: STREM CHEMICALS INC MS
Warning

38-2250-10G 38-2250-50G
100203-106EA 151.45 USD
100203-106 100200-676
Strontium oxide ≥99.9%
Strontium oxide
Formula: SrO
Boiling Pt: 3000 °C (1013 hPa)
Melting Pt: 2430 °C
Density: 4.7
MDL Number: MFCD00049558
CAS Number: 1314-11-0
EINECS: 215-219-9
UN: 3262
ADR: 8,II
Merck Index: 12,09009

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR