Mercury(II) bromide ACS

Supplier: STREM CHEMICALS INC MS
Danger

93-8003-250G 93-8003-50G
100208-600EA 745.91 USD
100208-600 100200-608
Mercury(II) bromide ACS
Mercury (II) bromide
Formula: Br₂Hg
Boiling Pt: 322 °C
Melting Pt: 236 °C
Density: 6.109
Storage Temperature: Ambient
MDL Number: MFCD00011040
CAS Number: 7789-47-1
EINECS: 232-169-3
UN: 1634
ADR: 6.1,II
Merck Index: 13,05900

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR