Diammonium tetrachloropalladate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Ammonium tetrachloropalladate(II, Ammonium tetrachloropalladate(II), Ammonium palladium (II) chloride, Ammonium tetrachloropalladate (II)

93-4602-1G 93-4602-5G
100200-576EA 113.23 USD
100200-576 100205-582
Diammonium tetrachloropalladate ≥99%
Diammonium tetrachloropalladate
Formula: H₈Cl₄N₂Pd
Melting Pt: Dec.
Density: 2,17
MDL Number: MFCD00010884
CAS Number: 13820-40-1
EINECS: 237-498-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR