Strontium chloride, anhydrous ≥95%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-3806-1000G
93-3806-250G
100200-136EA
281.66
USD
100200-136
100200-556
Strontium chloride, anhydrous ≥95%
Strontium chloride
Formula:
Cl₂Sr MW: 158.53 Boiling Pt: 1250 Melting Pt: 868 Density: 3.052 |
MDL Number:
MFCD00011249 CAS Number: 10476-85-4 EINECS: 233-971-6 Merck Index: 12,09000 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...