Copper(II) gluconate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Copper(II) (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate, Gluconic Acid Copper(II) Salt, Copper(II) D-gluconate, Copper Gluconate

93-2927-100G 93-2927-500G
100196-596EA 63.69 USD
100196-596 100200-550
Copper(II) gluconate ≥98%
Copper(II) gluconate
Formula: C₁₂H₂₂CuO₁₄
MDL Number: MFCD00075297
CAS Number: 527-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR