Ammonium hexafluorophosphate ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

93-1502-100G 93-1502-25G
100197-712EA 240.62 USD
100197-712 100200-518
Ammonium hexafluorophosphate ≥99%
Ammonium hexafluorophosphate
Formula: H₄F₆NP
Melting Pt: Dec.
Density: 2.180
MDL Number: MFCD00064642
CAS Number: 16941-11-0
EINECS: 241-009-1
UN: 3260
ADR: 8,II
Merck Index: 12,00557

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR