Tetraammineplatinum(II) nitrate ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

78-2010-5G 78-2010-250MG 78-2010-1G
100200-496EA 891.69 USD
100200-496 101442-472 100200-494
Tetraammineplatinum(II) nitrate ≥99%
Tetraammineplatinum(II) nitrate
Formula: H₁₂N₆O₆Pt
MDL Number: MFCD00011624
CAS Number: 20634-12-2
UN: 1477
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR