Ethyldiphenylphosphine ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

15-2150-5G 15-2150-25G
100200-430EA 86.34 USD
100200-430 100200-428
Ethyldiphenylphosphine ≥99%
Ethyldiphenylphosphine

Phosphine

Formula: C₁₄H₁₅P
Boiling Pt: 130…132 °C (1 mmHg)
Density: 1.04
Flash Pt: >230 °F
MDL Number: MFCD00015170
CAS Number: 607-01-2
UN: 3334
ADR: 9,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR