(±)-6,6'-Dibromo-1,1'-bi-2-naphthol ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: racemic-6,6'-Dibromo-1,1'-bi-2-naphthol, 6,6'-Dibromo[1,1'-binaphthalene]-2,2'-diol

08-0604-1G 08-0604-5G
100200-386EA 94.83 USD
100200-386 100208-280
(±)-6,6'-Dibromo-1,1'-bi-2-naphthol ≥98%
(±)-6,6'-Dibromo-1,1'-bi-2-naphthol
Formula: C₂₀H₁₂Br₂O₂
Melting Pt: 202…205 °C
MDL Number: MFCD00798290
CAS Number: 13185-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR