Bis(pinacolato)diboron ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 4,4,4',4',5,5,5',5'-Octamethyl-2,2'-bi(1,3,2-dioxaborolane), 4,4,5,5-tetramethyl-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane

05-0030-500MG 05-0030-2G
100200-370EA 39.63 USD
100200-370 100203-688
Bis(pinacolato)diboron ≥99%
Bis(pinacolato)diboron

Boronic Acid and Ester

Formula: C₁₂H₂₄B₂O₄
Melting Pt: 144…145 °C
Density: 0,98
MDL Number: MFCD00799570
CAS Number: 73183-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR