Cyclopentadienylindium(I) ≥99.99% (In basis), PURATREM, Electronic Grade
Supplier: STREM CHEMICALS INC MS
|
Danger
|
97-3425-5G
97-3425-1G
100203-642EA
2427.38
USD
100203-642
100200-358
Cyclopentadienylindium(I) ≥99.99% (In basis), PURATREM, Electronic Grade
Cyclopentadienylindium(I)
Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD
Boiling Pt:
50 °C (0.01 mmHg) (subl.) |
MDL Number:
MFCD00151371 CAS Number: 34822-89-4 UN: 1325 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...