Holmium(III) chloride, anhydrous ≥99.9% (Ho basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Holmium trichloride

93-6721-25G 93-6721-5G
100210-894EA 564.74 USD
100210-894 100200-190
Holmium(III) chloride, anhydrous ≥99.9% (Ho basis)
Holmium(III) chloride
Formula: Cl₃Ho
Boiling Pt: 1500 °C (1013 hPa)
Melting Pt: 718 °C
Density: 3.7 g/cm³ (20 °C)
MDL Number: MFCD00011051
CAS Number: 10138-62-2
EINECS: 233-387-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR