Iron(III) nitrate nonahydrate ≥99.999% (Fe basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Ferric nitrate nonahydrate, Iron trinitrate nonahydrate, Iron(III) nitrate hydrate, Ferric Nitrate, 9-Hydrate
93-2672-25G
93-2672-5G
100200-092EA
564.74
USD
100200-092
100206-984
Iron(III) nitrate nonahydrate ≥99.999% (Fe basis), PURATREM
Iron (III) nitrate nonahydrate
Formula:
Fe(NO₃)₃·9H₂O MW: 404 g/mol Boiling Pt: 125 (dec.) Melting Pt: 47 Density: 1.684 Storage Temperature: Ambient |
MDL Number:
MFCD00149708 CAS Number: 7782-61-8 EINECS: 233-899-5 UN: 1466 ADR: 5.1 III Merck Index: 13,04057 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...