Chromium(II) chloride, anhydrous ≥99.9% (Cr basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Chromium dichloride

93-2405-25G 93-2405-5G
100200-076EA 468.49 USD
100200-076 100203-418
Chromium(II) chloride, anhydrous ≥99.9% (Cr basis)
Chromium (II) chloride
Formula: Cl₂Cr
Boiling Pt: 1302 °C (1013 hPa)
Melting Pt: 824 °C
Density: 2.878
Storage Temperature: Ambient
MDL Number: MFCD00010947
CAS Number: 10049-05-5
EINECS: 233-163-3
UN: 3260
ADR: 8,III
Merck Index: 13,02264

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR