Nitrosium hexafluoroantimonate ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Nitrosyl hexafluoroantimonate, Nitrosonium hexafluoroantimonate

93-0722-25G
100200-032EA 435.94 USD
100200-032
Nitrosium hexafluoroantimonate ≥97%
Nitrosium hexafluoroantimonate
Formula: F₆NOSb
MDL Number: MFCD00042550
CAS Number: 16941-06-3
UN: 2923
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR