Dilithium metasilicate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

93-0377-50G 93-0377-250G
100200-022EA 117.48 USD
100200-022 100208-074
Dilithium metasilicate ≥99%
di-Lithium metasilicate
Formula: Li₂O₃Si
Melting Pt: 1201 °C
Density: 2.52
MDL Number: MFCD00016179
CAS Number: 10102-24-6
EINECS: 233-270-5
Merck Index: 12,05568

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR