Ammonium sulfate ≥99% ACS

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: sulfuric acid diammonium salt, Diammonium sulfate, Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

93-0225-2000G 93-0225-500G
100208-984EA 181.17 USD
100208-984 100200-012
Ammonium sulfate ≥99% ACS
Ammonium sulphate
Formula: (NH₄)₂SO₄
Melting Pt: 235 °C (dec.)
Density: 1.769
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR