Anti-ASC Mouse Monoclonal Antibody [clone: O93E9]

Supplier: Biolegend
676502
10020-806EA 395.05 USD
10020-806
Anti-ASC Mouse Monoclonal Antibody [clone: O93E9]
Antibodies
Purified anti-ASC [O93E9]; Isotype: Mouse IgG1; Reactivity: Human; Apps: WB; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 0.5 - 2.0 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: ASC
Clonality: Monoclonal
Clone: O93E9
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR