Anti-Ki-67 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 16A8]

Supplier: Biolegend
652419 652420
10020-544EA 173 USD
10020-544 10020-546
Anti-Ki-67 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 16A8]
Antibodies
Alexa Fluor® 700 anti-mouse Ki-67 [16A8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ICFC, IF; Size: 100 μg

Each lot of this antibody is quality control tested by our Ki-67 staining protocol below. For flow cytometric staining, the suggested use of this reagent is 0.125 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Ki-67
Clonality: Monoclonal
Clone: 16A8
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR