Anti-CD8a Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: RPA-T8]

Supplier: Biolegend
301022 301025 301062
301022-BLEA 363.16 USD
301022-BL 301025-BL 10020-378
Anti-CD8a Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: RPA-T8]
Antibodies
Alexa Fluor® 647 anti-human CD8a [RPA-T8]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus, Pigtailed Macaque, Sooty Mangabey; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining using the μg size, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. For flow cytometric staining using the test sizes, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: RPA-T8
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee;Baboon;Cynomolgus;Rhesus;Pigtailed Macaque;Sooty Mangabey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR