Anti-CD71 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: OX-26]

Supplier: Biolegend
204410
10020-354EA 216.5 USD
10020-354
Anti-CD71 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: OX-26]
Antibodies
PE anti-rat CD71 [OX-26]; Isotype: Mouse IgG2a, κ; Reactivity: Rat; Apps: FC; Size: 200 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD71
Clonality: Monoclonal
Clone: OX-26
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR