Palladium(II) acetate ≥98% (99.9+%-Pd)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Palladium diacetate, Palladium (II) acetate
46-1780-1G
46-1780-5G
100205-152EA
145.78
USD
100205-152
100199-946
Palladium(II) acetate ≥98% (99.9+%-Pd)
Palladium (II) acetate
Formula:
C₄H₆O₄Pd Melting Pt: 205 °C |
MDL Number:
MFCD00012453 CAS Number: 3375-31-3 EINECS: 222-164-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...