Rhodium ≥99.95%, Powder

Supplier: STREM CHEMICALS INC MS
Warning

45-1872-500MG
100199-932EA 1672.98 USD
100199-932
Rhodium ≥99.95%, Powder
Rhodium
Formula: Rh
MW: 102.9 g/mol
Boiling Pt: 3727 °C
Melting Pt: 1966 °C
Density: 12,41
MDL Number: MFCD00011201
CAS Number: 7440-16-6
EINECS: 231-125-0
UN: 3089
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR