Ferrocene ≥98%

Supplier: STREM CHEMICALS INC MS

26-1699-2000G 26-1699-500G
100199-836EA 464.25 USD
100199-836 100201-554
Ferrocene ≥98%
Ferrocene

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₀H₁₀Fe
MW: 186.04 g/mol
Boiling Pt: 249
Melting Pt: 172…173 °C
Density: 1.49 g/cm³ (20 °C)
MDL Number: MFCD00001427
CAS Number: 102-54-5
EINECS: 203-039-3
UN: 1325
ADR: 4.1 II
Merck Index: 13,04070

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR