Bis(cyclopentadienyl)vanadium ≥95%, sublimated

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Vanadocene

23-0180-5G 23-0180-1G
100199-828EA 546.34 USD
100199-828 100199-826
Bis(cyclopentadienyl)vanadium ≥95%, sublimated
Bis(cyclopentadienyl)vanadium

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₀H₁₀V
Boiling Pt: 200 °C (0.1 mmHg) (subl.)
MDL Number: MFCD00058735
CAS Number: 1277-47-0
UN: 1325
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR