(R)-(+)-2-Diphenylphosphino-2'-methoxy-1,1'-binaphthyl ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (R)-(+)-MOP, (R)-(2'-Methoxy-[1,1'-binaphthalen]-2-yl)diphenylphosphine, (R)-(+)-2-(Diphenylphosphino)-2'-methoxy-1,1'-binaphthyl, (R)-MOP

15-1775-500MG 15-1775-100MG
100199-798EA 560.49 USD
100199-798 100199-796
(R)-(+)-2-Diphenylphosphino-2'-methoxy-1,1'-binaphthyl ≥99%
(R)-(+)-2-Diphenylphosphino-2'-methoxy-1,1'-binaphthyl

Phosphine

Formula: C₃₃H₂₅OP
Melting Pt: 177…179 °C
MDL Number: MFCD00269686
CAS Number: 145964-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR