2-Hydroxyphenylboronic acid pinacol ester ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

05-1042-1G 05-1042-5G
100199-700EA 151.45 USD
100199-700 100199-702
2-Hydroxyphenylboronic acid pinacol ester ≥97%
2-Hydroxyphenylboronic acid pinacol ester

Boronic Acid and Ester

Formula: C₁₂H₁₇BO₃
Boiling Pt: 100 °C (0.2 mmHg)
MDL Number: MFCD02093754
CAS Number: 269409-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR