Molecular sieve A4 (0.4 nm, 4 Å), beads -4+8

Supplier: STREM CHEMICALS INC MS
Warning

93-5415-1000G 93-5415-250G
100199-620EA 155.69 USD
100199-620 100196-752
Molecular sieve A4 (0.4 nm, 4 Å), beads -4+8

Molecular Sieve

MDL Number: MFCD03457537
CAS Number: 70955-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR