Molecular sieve A4 (0.4 nm, 4 Å), pellets 1/8''

Supplier: STREM CHEMICALS INC MS
Warning

93-5405-1000G 93-5405-250G
100199-602EA 161.35 USD
100199-602 100196-736
Molecular sieve A4 (0.4 nm, 4 Å), pellets 1/8''
Molecular sieve A4 (0.4 nm, 4 Å)
MDL Number: MFCD03457537
CAS Number: 70955-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR