Molecular sieve A4 (0.4 nm, 4 Å), Powder -600 mesh

Supplier: STREM CHEMICALS INC MS
Warning

93-5403-250G 93-5403-1000G
100196-734EA 66.52 USD
100196-734 100199-600
Molecular sieve A4 (0.4 nm, 4 Å), Powder -600 mesh
Molecular sieve A4 (0.4 nm, 4 Å)

Molecular Sieve

MDL Number: MFCD03457537
CAS Number: 70955-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR