Cadmium ≥99.999%, powder

Supplier: STREM CHEMICALS INC MS
Danger

93-4848-10G 93-4848-50G
100199-572EA 418.95 USD
100199-572 100207-478
Cadmium ≥99.999%, powder
Cadmium
Formula: Cd
Boiling Pt: 765 °C
Melting Pt: 320.9 °C
Density: 8.642
MDL Number: MFCD00010914
CAS Number: 7440-43-9
EINECS: 231-152-8
UN: 2570
ADR: 6.1,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR