Chromium ≥99.8%, chips

Supplier: STREM CHEMICALS INC MS

93-2457-250G 93-2457-50G
100199-526EA 161.35 USD
100199-526 100208-532
Chromium ≥99.8%, chips
Chromium
Formula: Cr
Boiling Pt: 2200 °C
Melting Pt: 1890 °C
Density: 7.20
Storage Temperature: Ambient
MDL Number: MFCD00010944
CAS Number: 7440-47-3
EINECS: 231-157-5
Merck Index: 12,02288

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR