Titanium(II) hydride ≥95% (99+%-Ti)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Titanium hydride, Titanium dihydride

93-2211-100G 93-2211-25G 93-2211-500G
100199-510EA 151.45 USD
100199-510 100204-230 100203-412
Titanium(II) hydride ≥95% (99+%-Ti)
Titanium (II) hydride
Formula: H₂Ti
Melting Pt: 450 °C (dec.)
Density: 3.9
MDL Number: MFCD00151301
CAS Number: 7704-98-5
EINECS: 231-726-8
UN: 1871
ADR: 4.1,II
Merck Index: 14,09473

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR