Lithium diethylamide ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

93-0317-5G 93-0317-25G
100205-234EA 89.17 USD
100205-234 100199-464
Lithium diethylamide ≥95%
Lithium diethylamide
MDL Number: MFCD00040489
CAS Number: 816-43-3
UN: 2846
ADR: 4.2,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR