Tantalum ≥99.95%, foil

Supplier: STREM CHEMICALS INC MS
Warning

73-0040-25X25MM 73-0040-100X100MM 73-0040-50X50MM
100209-378EA 268.92 USD
100209-378 100199-438 100199-440
Tantalum ≥99.95%, foil
Tantalum
Formula: Ta
MW: 180.95 g/mol
Boiling Pt: 5427
Melting Pt: 2996
Density: 16.6
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR