Bis(pentamethylcyclopentadienyl)iron(II) ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Bis(pentamethylcyclopentadienyl)iron, Decamethylferrocene

26-0400-5G 26-0400-1G
100199-370EA 628.43 USD
100199-370 100199-368
Bis(pentamethylcyclopentadienyl)iron(II) ≥99%
Bis(pentamethylcyclopentadienyl)iron (II)

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₂₀H₃₀Fe
Melting Pt: 298…300 °C (subl.)
MDL Number: MFCD00058736
CAS Number: 12126-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR