2,3-Dimethyl-1-propylimidazolium bis(trifluoromethylsulfonyl)imide ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

07-0465-5G
100199-318EA 733.17 USD
100199-318
2,3-Dimethyl-1-propylimidazolium bis(trifluoromethylsulfonyl)imide ≥99%
2,3-Dimethyl-1-propylimidazolium bis(trifluoromethylsulphonyl)imide

Ionic Liquids

Formula: C₁₀H₁₅F₆N₃O₄S₂
Density: 1,47
MDL Number: MFCD03788922
CAS Number: 169051-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR