Titanium ≥99.99%, wire

Supplier: STREM CHEMICALS INC MS
Warning

93-2255-50CM 93-2255-250CM
100201-986EA 147.2 USD
100201-986 100199-274
Titanium ≥99.99%, wire
Titanium
Formula: Ti
Boiling Pt: 3262 °C (1013 hPa)
Melting Pt: 1660 °C
Density: 4.5
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR