Diethyl diselenide ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

34-0380-10G 34-0380-50G
100203-102EA 488.31 USD
100203-102 100199-234
Diethyl diselenide ≥97%
Diethyl diselenide

Metal Alkyls, Volatile Precursors for CVD

Formula: C₄H₁₀Se₂
Boiling Pt: 85 °C (21 mmHg)
MDL Number: MFCD00015491
CAS Number: 628-39-7
EINECS: 211-042-6
UN: 3440
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR