Pentamethylcyclopentadienylchromium dicarbonyl dimer ≥99%

Supplier: STREM CHEMICALS INC MS

24-0900-1G 24-0900-5G
100198-156EA 230.71 USD
100198-156 100199-230
Pentamethylcyclopentadienylchromium dicarbonyl dimer ≥99%
Pentamethylcyclopentadienylchromium dicarbonyl dimer

Metallocenes, Derivatives & Cp Precursors

Melting Pt: 200 °C (dec.)
MDL Number: MFCD00058843
CAS Number: 37299-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR