Silicon dioxide ≥99.999% (Si basis), PURATREM, Electronic Grade

Supplier: STREM CHEMICALS INC MS

98-0145-100G 98-0145-5G 98-0145-25G
100201-294EA 495.38 USD
100201-294 100199-214 100205-810
Silicon dioxide ≥99.999% (Si basis), PURATREM, Electronic Grade
Silicium dioxide
Formula: SiO₂
Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1500 °C
Density: 2.6
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 7631-86-9
EINECS: 231-545-4
Merck Index: 14,08493

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR