Bismuth(III) oxide ≥99.999% (Bi basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Bismuth Trioxide, Dibismuth trioxide

93-8319-500G 93-8319-100G 93-8319-25G
100199-204EA 673.72 USD
100199-204 100205-800 100196-842
Bismuth(III) oxide ≥99.999% (Bi basis), PURATREM
Bismuth (III) oxide
Formula: BiO₃
Boiling Pt: 1890 °C
Melting Pt: 817 °C
Density: 8.9
MDL Number: MFCD00003462
CAS Number: 1304-76-3
EINECS: 215-134-7
Merck Index: 13,01273

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR