Sodium tetrachloroplatinate(II) hydrate

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Disodium tetrachloroplatinate hydrate, Sodium platinum chloride hydrate, Sodium chloroplatinate(II) hydrate

93-7810-5G 93-7810-1G
100207-316EA 632.68 USD
100207-316 100199-190
Sodium tetrachloroplatinate(II) hydrate
Sodium tetrachloroplatinate(II) hydrate
Formula: Na₂PtCl₄·xH₂O
Melting Pt: 100 °C
MDL Number: MFCD00150188
CAS Number: 207683-21-4
EINECS: 233-051-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR