Ytterbium (III) sulfate octahydrate ≥99.9% (Yb basis)

Supplier: STREM CHEMICALS INC MS

93-7027-5G 93-7027-25G
100199-170EA 48.12 USD
100199-170 100202-274
Ytterbium (III) sulfate octahydrate ≥99.9% (Yb basis)
Ytterbium (III) sulphate octahydrate
Formula: O₁₂S₃Yb₂·8H₂O
Density: 3,286
MDL Number: MFCD00149887
CAS Number: 10034-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR