Holmium ≥99.9%, foil

Supplier: STREM CHEMICALS INC MS

93-6734-25X25MM 93-6734-50X50MM
100199-164EA 373.66 USD
100199-164 100207-142
Holmium ≥99.9%, foil
Holmium
Formula: Ho
Boiling Pt: 2695 °C
Melting Pt: 1474 °C
Density: 8,795
MDL Number: MFCD00011049
CAS Number: 7440-60-0
EINECS: 231-169-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR